Merck
Nanogel-based delivery of mycophenolic acid ameliorates systemic lupus erythematosus in mice.
The Journal of clinical investigation (2013-03-05)
Michael Look, Eric Stern, Qin A Wang, Leah D DiPlacido, Michael Kashgarian, Joe Craft, Tarek M Fahmy
ZUSAMMENFASSUNG
The ability to selectively inactivate immune cells with immunosuppressants is a much sought-after modality for the treatment of systemic lupus erythematosus and autoimmunity in general. Here, we designed and tested a novel nanogel drug delivery vehicle for the immunosuppressant mycophenolic acid (MPA). Treatment with MPA-loaded nanogels increased the median survival time (MST) of lupus-prone NZB/W F1 mice by 3 months with prophylactic use (MST was 50 weeks versus 38 weeks without treatment), and by 2 months when administered after the development of severe renal damage (MST after proteinuria onset was 12.5 weeks versus 4 weeks without treatment). Equivalent and greater doses of MPA administered in buffer were not efficacious. Nanogels had enhanced biodistribution to organs and association with immune cells. CD4-targeted nanogels yielded similar therapeutic results compared with nontargeted formulations, with protection from glomerulonephritis and decreases in IFN-γ-positive CD4 T cells. DCs that internalized nanogels helped mediate immunosuppression, as they had reduced production of inflammatory cytokines such as IFN-γ and IL-12. Our results demonstrate efficacy of nanogel-based lupus therapy and implicate a mechanism by which immunosuppression is enhanced, in part, by the targeting of antigen-presenting cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Polymilchsäure, Mw ~60,000
Sigma-Aldrich
Mycophenolsäure, ≥98%
Sigma-Aldrich
Mycophenolsäure, powder, BioReagent, suitable for cell culture
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®