Merck
  • Lipid management in 13,000 high risk cardiovascular patients treated under daily practice conditions: LIMA Registry.
Lipid management in 13,000 high risk cardiovascular patients treated under daily practice conditions: LIMA Registry.
Vascular health and risk management (2013-03-06)
Juergen R Schaefer, Anselm K Gitt, Frank Sonntag, Achim Weizel, Christina Jannowitz, Barbara Karmann, David Pittrow, Kurt Bestehorn
ZUSAMMENFASSUNG
We aimed to document the drug management of patients at high cardiovascular risk in daily practice, with the special focus on lipid-lowering treatment. In this prospective noninterventional study in 2387 outpatient centers throughout Germany, a total of 13,942 high-risk patients (mean age 65.7 years, 61.6% males) were treated with simvastatin 40 mg/day at entry as monotherapy. All patients were followed up for 12 months in terms of drug utilization, laboratory values, target attainment, and clinical events (including death, hospitalization, vascular events, and dialysis). Patients had coronary heart disease in 35.0%, diabetes mellitus in 24.4%, and the combination of coronary heart disease plus diabetes mellitus in 25.7%. In 21% of patients, a cholesterol absorption inhibitor was added to statin therapy at the entry visit, and in 23%, this was added at the follow up visit 6 months later. The target values for low-density lipoprotein-cholesterol (<2.6 mmol/L) were reached by 31.8% of patients at entry and by 50.0% at the end of this registry after 12 months. Mean blood pressure decreased (from 135.9/80.5 mmHg at baseline) by 3.1/1.9 mmHg after 12 months. In patients with documented diabetes, the targeted glycated hemoglobin (HbA1c <6.5%) was reached by 33.5% at baseline and by 40.0% after 12 months. Clinical events occurred in 11.7% of patients between baseline and month 6, and in 12.0% between months 6 and 12. In patients at high risk for cardiovascular events, comprehensive management under daily practice conditions leads to improvement of lipid, glucose, and blood pressure parameters. There is a need to improve secondary prevention among high-risk patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Simvastatin, European Pharmacopoeia (EP) Reference Standard