- Dose dependent stuttering with clozapine: a case report.
Dose dependent stuttering with clozapine: a case report.
Asian journal of psychiatry (2013-03-08)
Tarun Kumar, Archana Kathpal, Carrol Tim Longshore
PMID23466117
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Clozapin, European Pharmacopoeia (EP) Reference Standard
Clozapin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard