Merck
  • The renal angiotensin system does not reveal its secrets with a cross-sectional analysis of urinary angiotensinogen levels.
The renal angiotensin system does not reveal its secrets with a cross-sectional analysis of urinary angiotensinogen levels.
Journal of hypertension (2013-03-08)
Branko Braam
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Angiotensinogen aus Humanplasma, ≥80% (SDS-PAGE), lyophilized powder