Merck
  • Irreversible renal damage after transient renin-angiotensin system stimulation: involvement of an AT1-receptor mediated immune response.
Irreversible renal damage after transient renin-angiotensin system stimulation: involvement of an AT1-receptor mediated immune response.
PloS one (2013-03-08)
Bart F J Heijnen, Jelly Nelissen, Helma van Essen, Gregorio E Fazzi, Jan W Cohen Tervaert, Carine J Peutz-Kootstra, John J Mullins, Casper G Schalkwijk, Ben J A Janssen, Harry A J Struijker-Boudier
ZUSAMMENFASSUNG
Transient activation of the renin-angiotensin system (RAS) induces irreversible renal damage causing sustained elevation in blood pressure (BP) in Cyp1a1-Ren2 transgenic rats. In our current study we hypothesized that activation of the AT1-receptor (AT1R) leads to a T-cell response causing irreversible impairment of renal function and hypertension. Cyp1a1-Ren2 rats harbor a construct for activation of the RAS by indole-3-carbinol (I3C). Rats were fed a I3C diet between 4-8 weeks of age to induce hypertension. Next, I3C was withdrawn and rats were followed-up for another 12 weeks. Additional groups received losartan (20 mg/kg/day) or hydralazine (100 mg/kg/day) treatment between 4-8 weeks. Rats were placed for 24h in metabolic cages before determining BP at week 8, 12 and 20. At these ages, subsets of animals were sacrificed and the presence of kidney T-cell subpopulations was investigated by immunohistochemistry and molecular marker analysis. The development of sustained hypertension was completely prevented by losartan, whereas hydralazine only caused a partial decrease in BP. Markers of renal damage: KIM-1 and osteopontin were highly expressed in urine and kidney samples of I3C-treated rats, even until 20 weeks of age. Additionally, renal expression of regulatory-T cells (Tregs) was highly increased in I3C-treated rats, whereas the expression of T-helper 1 (Th1) cells demonstrated a strong decrease. Losartan prevented these effects completely, whereas hydralazine was unable to affect these changes. In young Cyp1a1-Ren2 rats AT1R activation leads to induction of an immune response, causing a shift from Th1-cells to Tregs, contributing to the development of irreversible renal damage and hypertension.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Losartan-Kalium
Sigma-Aldrich
Hydralazin -hydrochlorid
Losartan-Kalium, European Pharmacopoeia (EP) Reference Standard