Merck
  • Characterization of functionalized polyolefins by high-temperature two-dimensional liquid chromatography.
Characterization of functionalized polyolefins by high-temperature two-dimensional liquid chromatography.
Journal of chromatography. A (2013-03-12)
Anton Ginzburg, Tibor Macko, Frank Malz, Michael Schroers, Irene Troetsch-Schaller, Jan Strittmatter, Robert Brüll
ZUSAMMENFASSUNG
High-temperature two-dimensional liquid chromatography (HT 2D-LC) was developed for the separation and characterization of functional polyolefins. Therefore, the key experimental parameters, namely the injection volume, the mobile phase composition, the flow rate in SEC and the time and phase of sampling into the second dimension, were systematically varied and their influence on the resolution of separation were studied. The HPLC separation of ethylene-vinylacetate waxes was realized using silica gel as stationary phase and a solvent gradient decalin→cyclohexanone, while SEC separations were realized in the chromatographic system polystyrene divinyl benzene column/1,2,4-trichlorobenzene. By choosing suitable experimental parameters, the run time needed for one complete 2D-LC analysis of a polymer sample was shortened from about 200 min to 100 min. However, the developed method failed to adsorb polypropylene and ethylene/1-butene copolymers grafted with 13 or 3 mol.% of methyl methacrylate respectively. Using porous graphite as a stationary phase and a solvent gradient 1-decanol→1,2,4-trichlorobenzene as mobile phase 2D-LC separations of both grafted polyolefins were realized.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(methylmethacrylat), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methylmethacrylat)
Sigma-Aldrich
Poly(methylmethacrylat), average Mw ~15,000 by GPC, powder
Sigma-Aldrich
Poly(methylmethacrylat)
Sigma-Aldrich
Poly(ethylen-co-vinylacetat), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Sigma-Aldrich
Poly(ethylen-co-vinylacetat), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Poly(ethylen-co-vinylacetat), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Sigma-Aldrich
Poly(methylmethacrylat), isotaktisch, >80% isotactic
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methylmethacrylat), analytical standard, 2,480,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 8,000