- A transthylakoid proton gradient and inhibitors induce a non-photochemical fluorescence quenching in unicellular algae Nannochloropsis sp.
A transthylakoid proton gradient and inhibitors induce a non-photochemical fluorescence quenching in unicellular algae Nannochloropsis sp.
FEBS letters (2013-03-12)
Shaona Cao, Xiaowen Zhang, Dong Xu, Xiao Fan, Shanli Mou, Yitao Wang, Naihao Ye, Wenqi Wang
PMID23474242
ZUSAMMENFASSUNG
Non-photochemical quenching (NPQ) of chlorophyll fluorescence is thought to be an indicator of an essential regulation and photoprotection mechanism against high-light stress in photosynthetic organisms. In this report, special chemicals were used to perturb the kinetics of the ΔpH build-up and the xanthophyll cycle (XC) in Nannochloropsis sp. We found that NPQ was stimulated rapidly on exposure to high light and relaxed rapidly in darkness. The ΔpH could be obligatory for NPQ and ΔpH alone was not sufficient to induce NPQ. The XC, being strictly mediated by ΔpH, was also essential for NPQ. The results demonstrate that the mechanism of NPQ in Nannochloropsis sp. resembled that of diatoms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ammonium-Ionenlösung für ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)