Merck
  • [Regularity of changes in main active components of Schisandra chinensis before and after being processed with vinegar].
[Regularity of changes in main active components of Schisandra chinensis before and after being processed with vinegar].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-03-13)
Li Li, Yong-Qing Xiao, Ding-Rong Yu, Yin-Lian Ma, Ming-Gui Zhu, Liang Chen
ZUSAMMENFASSUNG
To investigate the regularity of changes in the main active components of Schisandra chinensis preparations before and after being processed with vinegar. High performance liquid chromatography was used to analyze the changes in the content of lignans and organic acids in S. chinensis preparations before and after being processed with vinegar. After being processed with vinegar, the content of lignans in S. chinensis preparations significantly reduced, while that of organic acids significantly increased. The ratio between them reduced from 1:16 (raw product) to 1:21 (vinegar products). The changes in the main pharmacological effects of S. chinensis are related to not only the changes in the contents of lignan and organic acids, but also the significant changes in the ratio relations between the two components. This is of great significance to the correlation among the principle of S. chinensis processed with vinegar and material basis and the changes in pharmacological effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Millipore
Bifido Selektives Supplement B, suitable for microbiology