Merck
A simple all-solid tellurite microstructured optical fiber.
Optics express (2013-03-14)
Tonglei Cheng, Zhongchao Duan, Meisong Liao, Weiqing Gao, Dinghuan Deng, Takenobu Suzuki, Yasutake Ohishi
ZUSAMMENFASSUNG
A simple all-solid tellurite microstructured optical fiber which has only one layer of high-index rods in the cladding is proposed and fabricated in the paper. The core and the cladding with the low index are made from the TeO(2)-ZnO-Na(2)O-La(2)O(3) glass, and the high-index rods are made from the TeO(2)-Li(2)O-WO(3)-MoO(3)-Nb(2)O(5) glass. The guiding regime in this fiber can be explained by ARROW model. The fiber can support the near- and mid-infrared light transmitting in the core within the transmission bands while the all-solid silica microstructured optical fiber cannot. When the pump light is outside the transmission bands, the light will transmit in six TLWMN rods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tellur, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellur, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellur, 99.999% trace metals basis
Sigma-Aldrich
Tellur, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellur, granular, −5-+50 mesh, 99.99% trace metals basis