Merck
  • Generation of stable human cell lines with Tetracycline-inducible (Tet-on) shRNA or cDNA expression.
Generation of stable human cell lines with Tetracycline-inducible (Tet-on) shRNA or cDNA expression.
Journal of visualized experiments : JoVE (2013-03-15)
Marta Gomez-Martinez, Debora Schmitz, Alexander Hergovich
ZUSAMMENFASSUNG
A major approach in the field of mammalian cell biology is the manipulation of the expression of genes of interest in selected cell lines, with the aim to reveal one or several of the gene's function(s) using transient/stable overexpression or knockdown of the gene of interest. Unfortunately, for various cell biological investigations this approach is unsuitable when manipulations of gene expression result in cell growth/proliferation defects or unwanted cell differentiation. Therefore, researchers have adapted the Tetracycline repressor protein (TetR), taken from the E. coli tetracycline resistance operon(1), to generate very efficient and tight regulatory systems to express cDNAs in mammalian cells(2,3). In short, TetR has been modified to either (1) block initiation of transcription by binding to the Tet-operator (TO) in the promoter region upon addition of tetracycline (termed Tet-off system) or (2) bind to the TO in the absence of tetracycline (termed Tet-on system) (Figure 1). Given the inconvenience that the Tet-off system requires the continuous presence of tetracycline (which has a half-life of about 24 hr in tissue cell culture medium) the Tet-on system has been more extensively optimized, resulting in the development of very tight and efficient vector systems for cDNA expression as used here. Shortly after establishment of RNA interference (RNAi) for gene knockdown in mammalian cells(4), vectors expressing short-hairpin RNAs (shRNAs) were described that function very similar to siRNAs(5-11). However, these shRNA-mediated knockdown approaches have the same limitation as conventional knockout strategies, since stable depletion is not feasible when gene targets are essential for cellular survival. To overcome this limitation, van de Wetering et al.(12) modified the shRNA expression vector pSUPER(5) by inserting a TO in the promoter region, which enabled them to generate stable cell lines with tetracycline-inducible depletion of their target genes of interest. Here, we describe a method to efficiently generate stable human Tet-on cell lines that reliably drive either inducible overexpression or depletion of the gene of interest. Using this method, we have successfully generated Tet-on cell lines which significantly facilitated the analysis of the MST/hMOB/NDR cascade in centrosome(13,14) and apoptosis signaling(15,16). In this report, we describe our vectors of choice, in addition to describing the two consecutive manipulation steps that are necessary to efficiently generate human Tet-on cell lines (Figure 2). Moreover, besides outlining a protocol for the generation of human Tet-on cell lines, we will discuss critical aspects regarding the technical procedures and the characterization of Tet-on cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetracyclin -hydrochlorid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracyclin -hydrochlorid, ≥95% (European Pharmacopoeia HPLC assay), meets EP testing specifications
Sigma-Aldrich
Tetracyclin -hydrochlorid, powder
Supelco
Tetracyclin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracyclin -hydrochlorid, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracyclin -hydrochlorid, meets USP testing specifications