Merck
  • Toward improved understanding of a potential association between isotretinoin and inflammatory bowel disease.
Toward improved understanding of a potential association between isotretinoin and inflammatory bowel disease.
The Journal of investigative dermatology (2013-03-15)
Alisa N Femia, Ruth Ann Vleugels
ZUSAMMENFASSUNG
Isotretinoin is a widely prescribed medication for nodulocystic acne. Existing literature is conflicting regarding an association between isotretinoin and inflammatory bowel disease. In this issue, Alhusayen et al. report findings from a 12-year observational study exploring a possible association, and they conclude that acne itself may be responsible for an apparent correlation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
13-cis-Retinolsäure, ≥98% (HPLC)
Supelco
Isotretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Isotretinoin, European Pharmacopoeia (EP) Reference Standard