Merck
  • Plants as bio-indicators of subsurface conditions: impact of groundwater level on BTEX concentrations in trees.
Plants as bio-indicators of subsurface conditions: impact of groundwater level on BTEX concentrations in trees.
International journal of phytoremediation (2013-03-16)
Jordan Wilson, Rachel Bartz, Matt Limmer, Joel Burken
ZUSAMMENFASSUNG
Numerous studies have demonstrated trees' ability to extract and translocate moderately hydrophobic contaminants, and sampling trees for compounds such as BTEX can help delineate plumes in the field. However, when BTEX is detected in the groundwater, detection in nearby trees is not as reliable an indicator of subsurface contamination as other compounds such as chlorinated solvents. Aerobic rhizospheric and bulk soil degradation is a potential explanation for the observed variability of BTEX in trees as compared to groundwater concentrations. The goal of this study was to determine the effect of groundwater level on BTEX concentrations in tree tissue. The central hypothesis was increased vadose zone thickness promotes biodegradation of BTEX leading to lower BTEX concentrations in overlying trees. Storage methods for tree core samples were also investigated as a possible reason for tree cores revealing lower than expected BTEX levels in some sampling efforts. The water level hypothesis was supported in a greenhouse study, where water table level was found to significantly affect tree BTEX concentrations, indicating that the influx of oxygen coupled with the presence of the tree facilitates aerobic biodegradation of BTEX in the vadose zone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Ethylbenzol, ReagentPlus®, 99%
Supelco
Benzol -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, analytical standard
Sigma-Aldrich
Ethylbenzol, anhydrous, 99.8%
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylbenzol, analytical standard
Supelco
Toluol -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D)
Supelco
Ethylbenzol -Lösung, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.