- Molecular dynamic simulation of mGluR5 amino terminal domain: essential dynamics analysis captures the agonist or antagonist behaviour of ligands.
Molecular dynamic simulation of mGluR5 amino terminal domain: essential dynamics analysis captures the agonist or antagonist behaviour of ligands.
Journal of molecular graphics & modelling (2013-03-19)
Alessandro Casoni, Francesca Clerici, Alessandro Contini
PMID23500630
ZUSAMMENFASSUNG
We describe the application of molecular dynamics followed by principal component analysis to study the inter-domain movements of the ligand binding domain (LBD) of mGluR5 in response to the binding of selected agonists or antagonists. Our results suggest that the method is an attractive alternative to current approaches to predict the agonist-induced or antagonist-blocked LBD responses. The ratio between the eigenvalues of the first and second eigenvectors (R1,2) is also proposed as a numerical descriptor for discriminating the ligand behavior as a mGluR5 agonist or antagonist.
MATERIALIEN