Merck
  • Anesthetic efficacy of a combination of 0.5 M mannitol plus 127.2 mg of lidocaine with 50 μg epinephrine in inferior alveolar nerve blocks: a prospective randomized, single-blind study.
Anesthetic efficacy of a combination of 0.5 M mannitol plus 127.2 mg of lidocaine with 50 μg epinephrine in inferior alveolar nerve blocks: a prospective randomized, single-blind study.
Anesthesia progress (2013-03-20)
Steven Smith, Al Reader, Melissa Drum, John Nusstein, Mike Beck
ZUSAMMENFASSUNG
The purpose of this prospective, randomized, single-blind study was to determine the anesthetic efficacy of 127.2 mg lidocaine with 50 μg epinephrine compared to 127.2 mg lidocaine with 50 μg epinephrine plus 0.5 M mannitol in inferior alveolar nerve (IAN) blocks. Forty subjects randomly received 2 IAN blocks consisting of a 3.18 mL formulation of 127.2 mg lidocaine with 50 μg epinephrine and a 5 mL formulation of 127.2 mg lidocaine with 50 μg epinephrine (3.18 mL) plus 0.5 M mannitol (1.82 mL) in 2 separate appointments spaced at least 1 week apart. Mandibular anterior and posterior teeth were blindly electric pulp tested at 4-minute cycles for 60 minutes postinjection. Pain of solution deposition and postoperative pain were also measured. No response from the subject to the maximum output (80 reading) of the pulp tester was used as the criterion for pulpal anesthesia. Total percent pulpal anesthesia was defined as the total of all the times of pulpal anesthesia (80 readings) over the 60 minutes. One hundred percent of the subjects had profound lip numbness with both inferior alveolar nerve blocks. The results demonstrated that a 5 mL formulation of 127.2 mg lidocaine with 50 μg epinephrine plus 0.5 M mannitol was significantly better than the 3.18 mL formulation of 127.2 mg lidocaine with 50 μg epinephrine for all teeth. Solution deposition pain and postoperative pain were not statistically different between the lidocaine/mannitol formulation and the lidocaine formulation without mannitol. We concluded that adding 0.5 M mannitol to a lidocaine with epinephrine formulation was significantly more effective in achieving a greater percentage of total pulpal anesthesia than a lidocaine formulation without mannitol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination