- [Use of metabolic and pathogenetic criteria in group standardization of organically active chlortriazine dyes].
[Use of metabolic and pathogenetic criteria in group standardization of organically active chlortriazine dyes].
Gigiena truda i professional'nye zabolevaniia (1990-01-01)
B A Kurliandskiĭ, L A Bazarova, E L Balabanova, A M Kliachkina, L A Dement'eva, I V Pervukhina, T A Kasatkina, N P Parinova, N I Nevzorova
PMID2351281
ZUSAMMENFASSUNG
The article presents research data related to the biotransformation, intoxication mechanisms and pathogenic properties of chlortriazine dye-stuffs. All the dye-stuffs studies were characterized by similar intoxication properties and pathogenesis, and were related to the 3rd class of hazardous substances. The MAC for the organic chlortriazine dye-stuffs constituted 2 mg/m3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.