Merck
  • Heterologous expression, purification, crystallization and preliminary X-ray analysis of Trichoderma reesei xylanase II and four variants.
Heterologous expression, purification, crystallization and preliminary X-ray analysis of Trichoderma reesei xylanase II and four variants.
Acta crystallographica. Section F, Structural biology and crystallization communications (2013-03-23)
Qun Wan, Andrey Kovalevsky, Qiu Zhang, Scott Hamilton-Brehm, Rosalynd Upton, Kevin L Weiss, Marat Mustyakimov, David Graham, Leighton Coates, Paul Langan
ZUSAMMENFASSUNG
Xylanase II from Trichoderma reesei catalyzes the hydrolysis of glycosidic bonds in xylan. Crystallographic studies of this commercially important enzyme have been initiated to investigate its reaction mechanism, substrate binding and dependence on basic pH conditions. The wild-type protein was heterologously expressed in an Escherichia coli host using the defined medium and four active-site amino acids were replaced to abolish its activity (E177Q and E86Q) or to change its pH optimum (N44D and N44H). Cation-exchange and size-exclusion chromatography were used to obtain >90% protein purity. The ligand-free proteins and variant complexes containing substrate (xylohexaose) or product (xylotriose) were crystallized in several different space groups and diffracted to high resolutions (from 1.07 to 1.55 Å).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Xylanase aus Trichoderma viride, lyophilized powder, ≥100 units/mg protein