Merck
Effects of selenoprotein P on the contraction and relaxation of the airway smooth muscle.
General physiology and biophysics (2013-03-28)
Sachie K Ogawa, Min-Chul Shin, Masaki Hirashima, Norio Akaike, Yushi Ito
ZUSAMMENFASSUNG
Selenoprotein P (SeP) not only represents the major selenoprotein in plasma, but also provides more than 50% of the total plasma selenium. However, there is no report concerning the direct action of selenium or selenium-containing compounds on the contraction and relaxation of the airway smooth muscle. Therefore, we investigated the effects of SeP and sodium selenite (SS) on the indirectly induced contraction and relaxation of the cat bronchi, and gel contraction of cultured bovine tracheal smooth muscle cells (BTSMC) induced by ATP. In the present results, SeP or SS suppressed the amplitude of twitch-like contractions of cat bronchiole without affecting the non-adrenergic and non-cholinergic (NANC) relaxations evoked by electrical field stimulation. SeP also suppressed the ATP-induced gel contraction of BTSMC. These results suggest that SeP suppresses the amplitude of twitch-like contraction of cat bronchiole by acting directly on the bronchiolar smooth muscle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Natriumselenit, 99%
Sigma-Aldrich
Natriumselenit, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Natriumselenit, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Natriumselenit, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis