Merck
  • Excellence in exotics: case report: Encephalitozoon cuniculi-associated phacoclastic uveitis in a dwarf rabbit.
Excellence in exotics: case report: Encephalitozoon cuniculi-associated phacoclastic uveitis in a dwarf rabbit.
Compendium (Yardley, PA) (2012-01-01)
Anthony A Pilny
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fenbendazol, ≥98%
Supelco
Fenbendazol, VETRANAL®, analytical standard