- [Acute toloxatone poisoning. Apropos of 122 cases].
[Acute toloxatone poisoning. Apropos of 122 cases].
Therapie (1990-03-01)
P Azoyan, R Garnier, F J Baud, M L Efthymiou
PMID2353326
ZUSAMMENFASSUNG
Toloxatone is a new monoamine oxidase inhibitor. One hundred and twenty two cases of poisoning with this drug are reported. In this series, the minimal toxic dose was 2 g. The first symptoms appeared about one hour after ingestion. In most cases, only drowsiness and mild adrenergic effects were observed. In a few cases of massive overdose, coma, pyramidal irritation, and myoclonic jerks occurred. In 3 cases of severe poisoning, toloxatone was associated with tricyclic antidepressants. Symptoms were similar to those reported in intoxications associating classical monoamine oxidase inhibitors and tricyclic antidepressants: muscular rigidity, hyperthermia and cardiovascular collapse. Two of these patients died.
MATERIALIEN