Merck
  • Adherence challenges and long-acting injectable antipsychotic treatment in patients with schizophrenia.
Adherence challenges and long-acting injectable antipsychotic treatment in patients with schizophrenia.
Journal of psychosocial nursing and mental health services (2013-04-03)
N Kirk Morton, Donna Zubek
ZUSAMMENFASSUNG
Medication nonadherence has been associated with persistence of psychotic symptoms, relapse, and hospitalization in patients with schizophrenia. Patients with untreated psychosis are significantly less likely to achieve remission, whereas antipsychotic drug adherence has been associated with recovery. As such, adherence to antipsychotic drug treatment is a key issue for nurses and treatment team members caring for patients who typically are on chronic, progressive disease course. Long-acting injectable (LAI) anti-psychotic drugs, developed to improve adherence and provide and alternative antipsychotic drug treatment fro schizophrenia, have been associated with improved treatment outcomes including reduction of relapse rates approximately 30% and reduction in hospitalizations. However, LAI antipsychotic drugs remain underutilized in the United States despite a growing body of literature supporting positive outcomes of LAI versus oral antipsychotic drugs. Mental health nurses are in a key position to support improved adherence inpatients with schizophrenia through use of practical educational strategies that help patients, family members, and health care providers better understand and manage treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Risperidon, ≥98% (HPLC), powder
Supelco
Olanzapine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Olanzapin, ≥98% (HPLC)
Supelco
Haloperidol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Risperidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Fluphenazindecanoat, European Pharmacopoeia (EP) Reference Standard
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Risperidon für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Risperidon, European Pharmacopoeia (EP) Reference Standard