Merck
  • Break Out: urogenital schistosomiasis and Schistosoma haematobium infection in the post-genomic era.
Break Out: urogenital schistosomiasis and Schistosoma haematobium infection in the post-genomic era.
PLoS neglected tropical diseases (2013-04-05)
Paul J Brindley, Peter J Hotez
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Praziquantel, VETRANAL®, analytical standard