Merck
Protein chemical synthesis by serine and threonine ligation.
Proceedings of the National Academy of Sciences of the United States of America (2013-04-10)
Yinfeng Zhang, Ci Xu, Hiu Yung Lam, Chi Lung Lee, Xuechen Li
ZUSAMMENFASSUNG
An efficient method has been developed for the salicylaldehyde ester-mediated ligation of unprotected peptides at serine (Ser) or threonine (Thr) residues. The utility of this peptide ligation approach has been demonstrated through the convergent syntheses of two therapeutic peptides--ovine-corticoliberin and Forteo--and the human erythrocyte acylphosphatase protein (∼11 kDa). The requisite peptide salicylaldehyde ester precursor is prepared in an epimerization-free manner via Fmoc-solid-phase peptide synthesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Salicylaldehyd, reagent grade, 98%
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
SAFC
L-Threonin
Sigma-Aldrich
Salicylaldehyd, redist., ≥99.0% (GC)
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Humanes Parathyroid-Hormon-Fragment 1–34, ≥95% (HPLC), powder
Sigma-Aldrich
Salicylaldehyd, ≥98%, FG
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylaldehyd, analytical standard
Supelco
L-Threonin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)