Merck
  • Mechanistic understanding of tetracycline sorption on waste tire powder and its chars as affected by Cu(2+) and pH.
Mechanistic understanding of tetracycline sorption on waste tire powder and its chars as affected by Cu(2+) and pH.
Environmental pollution (Barking, Essex : 1987) (2013-04-17)
Fei Lian, Zhengguo Song, Zhongqi Liu, Lingyan Zhu, Baoshan Xing
ZUSAMMENFASSUNG
The sorption characteristics of tetracycline (TC) by waste tire powder and its chars were investigated to explore the potential of using waste tires as effective sorbents for removal of TC from aqueous solution. Naphthalene (NAPH), a typical hydrophobic organic compound, was selected as asorbate for comparison. TC displayed much lower sorption affinity to tire powder than NAPH. However, it exhibited similar adsorption affinity as NAPH on the pyrolyzed tire chars, which was mainly attributed to π-π electron-donor-acceptor interactions of TC with the graphite surface of chars. TC and Cu(2+) could mutually facilitate the sorption of each other on both tire powder and pyrolyzed chars in a wide pH range. This could be explained by the metallic complexation and/or surface-bridging mechanisms (i.e., Cu- or TC-bridging). However, Cu(2+) and NAPH depressed the sorption of each other on tire powder and displayed negligible impact to each other on the highly pyrolyzed chars.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetracyclin -hydrochlorid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Naphthalin, 99%
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracyclin -hydrochlorid, ≥95% (European Pharmacopoeia HPLC assay), meets EP testing specifications
Supelco
Naphthalin, analytical standard
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Supelco
Schmelzpunktstandard 79-81 °C, analytical standard
Sigma-Aldrich
Tetracyclin -hydrochlorid, powder
Supelco
Tetracyclin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naphthalin -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Tetracyclin -hydrochlorid, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracyclin -hydrochlorid, meets USP testing specifications
Supelco
Naphthalin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland