Merck
In vitro activity of the nisin dehydratase NisB.
Proceedings of the National Academy of Sciences of the United States of America (2013-04-17)
Neha Garg, Luis M A Salazar-Ocampo, Wilfred A van der Donk
ZUSAMMENFASSUNG
The biosynthesis of several classes of ribosomally synthesized and posttranslationally modified peptides involves dehydration of serine and threonine residues. For class I lantibiotics, thiopeptides, and goadsporin, this dehydration is catalyzed by lanthionine biosynthetic enzyme B (LanB) or LanB-like proteins. Although LanB proteins have been studied since 1992, in vitro reconstitution of their dehydration activity has been elusive. We show here the in vitro activity of the dehydratase involved in the biosynthesis of the food preservative nisin (NisB). In vitro, NisB dehydrated its substrate peptide NisA eight times in the presence of glutamate, ATP, Mg(2+), and the ribosomal/membrane fraction of bacterial cell extract. Mutation of 23 highly conserved residues of NisB identified a number of amino acids that are essential for dehydration activity. In addition, these mutagenesis studies identified three mutants, R786A, R826A, and H961A, that result in multiple glutamylations of the NisA substrate. Glutamylation was observed during both Escherichia coli coexpression of NisA with these mutants and in vitro assays. Treatment of the glutamylated substrate with WT NisB results in dehydrated NisA, suggesting that the glutamylated peptide is an intermediate in dehydration. Collectively, these studies suggest that dehydration involves glutamylation of the side chains of Ser and Thr followed by elimination. The latter step has precedent in the virginiamycin resistance protein virginiamycin B lyase. These studies will facilitate investigation of other LanB proteins involved in the biosynthesis of lantibiotics, thiopeptides, and goadsporin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Histidin, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidin, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Nisin aus Lactococcus lactis, potency: ≥900 IU/mg
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
SAFC
L-Histidin
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanin, BioUltra, ≥99.5% (NT)
Supelco
L-Histidin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanin, ≥99%
Supelco
L-Alanin, Pharmaceutical Secondary Standard; Certified Reference Material
Alanin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Histidin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanin-12C3, 99.9 atom % 12C