Merck
DPP-4 inhibitors improve cognition and brain mitochondrial function of insulin-resistant rats.
The Journal of endocrinology (2013-04-18)
Hiranya Pintana, Nattayaporn Apaijai, Nipon Chattipakorn, Siriporn C Chattipakorn
ZUSAMMENFASSUNG
Recent evidence has demonstrated that insulin resistance is related to the development of type 2 diabetes mellitus. Our previous study found that high-fat diet (HFD) consumption caused not only peripheral and brain insulin resistance but also brain mitochondrial dysfunction and cognitive impairment. Vildagliptin and sitagliptin, dipeptidyl-peptidase-4 inhibitors, are recently developed anti-diabetic drugs. However, the effects of both drugs on cognitive behaviors and brain mitochondrial function in HFD-induced insulin-resistant rats have not yet been investigated. Sixty male Wistar rats were divided into two groups to receive either normal diet or HFD for 12 weeks. Rats in each group were then further divided into three treatment groups to receive either vehicle, vildagliptin (3 mg/kg per day), or sitagliptin (30 mg/kg per day) for 21 days. The cognitive behaviors of the rats were tested using the Morris Water Maze test. Blood samples were collected to determine metabolic parameters and plasma oxidative stress levels. Upon completion of the study, the animals were killed and the brains were removed to investigate brain and hippocampal mitochondrial function as well as to determine oxidative stress levels. We demonstrated that both drugs significantly improved the metabolic parameters and decreased circulating and brain oxidative stress levels in HFD-induced insulin-resistant rats. In addition, both drugs completely prevented brain and hippocampal mitochondrial dysfunction and equally improved the learning behaviors impaired by the HFD. Our findings suggest that the inhibition of dipeptidyl-peptidase-4 enzymes with vildagliptin or sitagliptin in insulin-resistant rats not only increases peripheral insulin sensitivity but also decreases brain dysfunction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adamantan, ≥99%