Merck
  • Identification of selective and potent inhibitors of fibroblast activation protein and prolyl oligopeptidase.
Identification of selective and potent inhibitors of fibroblast activation protein and prolyl oligopeptidase.
Journal of medicinal chemistry (2013-04-19)
Sarah E Poplawski, Jack H Lai, Youhua Li, Zhiping Jin, Yuxin Liu, Wengen Wu, Yong Wu, Yuhong Zhou, James L Sudmeier, David G Sanford, William W Bachovchin
ZUSAMMENFASSUNG
Fibroblast activation protein (FAP) is a serine protease selectively expressed on reactive stromal fibroblasts of epithelial carcinomas. It is widely believed to play a role in tumor invasion and metastasis and therefore to represent a potential new drug target for cancer. Investigation into its biological function, however, has been hampered by the current unavailability of selective inhibitors. The challenge has been in identifying inhibitors that are selective for FAP over both the dipeptidyl peptidases (DPPs), with which it shares exopeptidase specificity, and prolyl oligopeptidase (PREP), with which it shares endopeptidase specificity. Here, we report the first potent FAP inhibitor with selectivity over both the DPPs and PREP, N-(pyridine-4-carbonyl)-d-Ala-boroPro (ARI-3099, 6). We also report a similarly potent and selective PREP inhibitor, N-(pyridine-3-carbonyl)-Val-boroPro (ARI-3531, 22). Both are boronic acid based inhibitors, demonstrating that high selectivity can be achieved using this electrophile. The inhibitors are stable, easy to synthesize, and should prove to be useful in helping to elucidate the biological functions of these two unique and interesting enzymes, as well as their potential as drug targets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Prolin
Sigma-Aldrich
L-Prolin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Prolin, 99%, FCC, FG
Supelco
L-Prolin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland