Merck
Non-syndromic tooth agenesis associated with a nonsense mutation in ectodysplasin-A (EDA).
Journal of dental research (2013-04-23)
T Nikopensius, T Annilo, T Jagomägi, C Gilissen, M Kals, K Krjutškov, R Mägi, M Eelmets, U Gerst-Talas, M Remm, M Saag, A Hoischen, A Metspalu
ZUSAMMENFASSUNG
Mutations in the ectodysplasin-A (EDA) gene have been generally associated with X-linked hypohidrotic ectodermal dysplasia (XLHED). Recently, missense mutations in EDA have been reported to cause familial non-syndromic tooth agenesis. In this study, we report a novel EDA mutation in an Estonian family segregating non-syndromic tooth agenesis with variable expressivity. Affected individuals had no associated defects in other ectodermal organs. Using whole-exome sequencing, we identified a heterozygous nonsense mutation c.874G>T (p.Glu292X) in the TNF homology domain of EDA in all affected female patients. This protein-altering variant arose de novo, and the potentially causative allele was transmitted to affected offspring from the affected mother. We suggest that the dental phenotype variability described in heterozygous female carriers of EDA mutation may occur because of the differential pattern of X-chromosome inactivation, which retains reduced levels of EDA-receptor signaling in tissues involved in tooth morphogenesis. This results in selective tooth agenesis rather than XLHED phenotype. The present study broadens the mutation spectrum for this locus and demonstrates that EDA mutations may result in non-syndromic tooth agenesis in heterozygous females.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutaminlösung 200 mmol/l, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Guanin, 98%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Thymin, ≥99%
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
Guanin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Guanin, BioUltra
Sigma-Aldrich
Thymin, BioReagent, suitable for cell culture
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland