Merck
  • Ferritin concentrations correlate to outcome of hematopoietic stem cell transplantation but do not serve as biomarker of graft-versus-host disease.
Ferritin concentrations correlate to outcome of hematopoietic stem cell transplantation but do not serve as biomarker of graft-versus-host disease.
Annals of hematology (2013-04-23)
M Großekatthöfer, E D Güclü, A Lawitschka, S Matthes-Martin, G Mann, M Minkov, C Peters, M G Seidel
ZUSAMMENFASSUNG
Clinical presentation and laboratory data are often too unspecific to distinguish the onset or activity of graft-versus-host disease (GvHD) from infections or toxicity. Antigen-presenting cells such as monocytes/macrophages and dendritic cells are involved in GvHD pathogenesis after allogeneic hematopoietic stem cell transplantation (HSCT). To test whether ferritin, an iron storage marker and macrophage activation-linked acute-phase protein, represents a candidate biomarker for acute or chronic GvHD in pediatric HSCT, we retrospectively evaluated a 2-year follow-up data from 131 eligible consecutive patients with different malignant and nonmalignant diseases who underwent allogeneic HSCT. Thirteen patients (10 %) suffered from acute GvHD II-IV°, 18 (14 %) had limited, and 14 (11 %) had extensive chronic GvHD. In extension of previous studies in adults investigating pre-transplant ferritin, our data show that post-HSCT hyperferritinemia (analyzed on days 0, +30, +60, +100, +180, +360, and +720) was significantly associated with decreased long-term survival (p < 0.001-0.03) in children and adolescents. Increased ferritin concentrations were associated with number and timing of red blood cell transfusions and toxic or infectious multi-organ failure but did not show significant differences between patients without GvHD and with acute grades II-IV, limited, or extensive chronic GvHD. Thus, our data do not identify ferritin as specifically GvHD-linked biomarker; however, they support the prognostic value of ferritin levels for outcome after HSCT in children.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ferritin aus Pferdemilz, Type I, saline solution
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide