Merck
  • A new simplified technique for making tricuspid expanded polytetrafluoroethylene valved conduit for right ventricular outflow reconstruction.
A new simplified technique for making tricuspid expanded polytetrafluoroethylene valved conduit for right ventricular outflow reconstruction.
The Annals of thoracic surgery (2013-04-24)
Hyungtae Kim, Si Chan Sung, Yun Hee Chang, Hyoung Doo Lee, Ji Ae Park
ZUSAMMENFASSUNG
Several methods of making expanded polytetrafluoroethylene valved conduit have been reported, but these techniques are complicated and less reproducible. We have developed a new simplified technique for making a tricuspid expanded polytetrafluoroethylene valved conduit for right ventricular outflow tract reconstruction and have been using this conduit for approximately 3 years with excellent results. Our technique is straightforward and reproducible, and the cusps in the conduit are durable due to the use of a specialized suturing technique.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polytetrafluorethylen, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), beads
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, ≥350 μm particle size