Merck
  • Mutualistic growth of the sulfate-reducer Desulfovibrio vulgaris Hildenborough with different carbohydrates.
Mutualistic growth of the sulfate-reducer Desulfovibrio vulgaris Hildenborough with different carbohydrates.
Mikrobiologiia (2013-04-25)
M M Santana, M C Portillo, J M Gonzalez
ZUSAMMENFASSUNG
Desulfovibrio vulgaris Hildenborough genome presents a phosphotransferase system putatively involved in the transport of carbohydrates. However, utilization of sugars by this sulfate-reducing bacterium has never been reported. Herein, we have observed proliferation of D. vulgaris Hildenborough with some carbohydrates, in mutualism with Stenotrophomonas maltophilia, a non-fermentative, gram-negative gammaproteobacterium, or Microbacterium, a gram-positive actinobacterium. These results suggest the importance of feedback interactions between different heterotrophic bacterial species including the alternative for D. vulgaris of exploiting additional organic resources and novel habitats. Thus, D. vulgaris strongly participates in the mineralization of carbohydrates both in complex natural and artificial systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
DL-Arabinose, ≥98% (GC)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination