Merck
  • Free radical scavenging and total antioxidant capacity of root extracts of Anchomanes difformis Engl. (Araceae).
Free radical scavenging and total antioxidant capacity of root extracts of Anchomanes difformis Engl. (Araceae).
Acta poloniae pharmaceutica (2013-04-25)
Abubakar B Aliyu, Mohammed A Ibrahim, Aliyu M Musa, Aisha O Musa, Joyce J Kiplimo, Adebayo O Oyewale
ZUSAMMENFASSUNG
Antioxidants activities from plants sources have attracted a wide range of interest across the world in recent times. This is due to growing concern for safe and alternative sources of antioxidants. The free radical scavenging activity using 1,1-diphenyl-2-picrylhydrazyl radical (DPPH), reducing power assay, total antioxidant capacity of the phosphomolybdenum method and the total phenolics content using the Folin-Ciocalteu reagent were carried out on the acetone, n-butanol and methanol root extracts of Anchomanes difformis. The results of the total phenolics content expressed in mg/100 g of gallic acid equivalent (GAE) showed that the n-butanol extract has significantly (p < 0.05) higher phenolics content (381 +/- 1.13) than the methanol and acetone extracts. All the extracts displayed strong concentration dependent radical scavenging activity. It was also observed that the n-butanol extract showed higher activity of 70.87% and 78.59% at low concentrations of 31.25 microg/mL and 62.5 microg/mL, respectively, than methanol and acetone extracts. The results also showed that the n-butanol extract has strongest reducing ability which is comparable to that of gallic acid at all the concentrations tested. Phytochemical screening on the extracts revealed the presence of flavonoids, saponins, and tannins. The results suggest that n-butanol extract of the plant is very rich in antioxidant compounds worthy of further investigations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Aceton, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Gallussäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Molybdän, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Butylalkohol, natural, ≥99.5%, FCC, FG