Merck
  • Determination of doxycycline in chicken fat by liquid chromatography with UV detection and liquid chromatography-tandem mass spectrometry.
Determination of doxycycline in chicken fat by liquid chromatography with UV detection and liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-04-25)
Anna Gajda, Andrzej Posyniak, Jan Zmudzki, Grzegorz Tomczyk
ZUSAMMENFASSUNG
A sensitive analytical method for determination of doxycycline (DC) residues in chicken fat/fat and skin was developed. The extraction, in the presence of the internal standard (IS) minocycline (MINO), was carried out using solution of oxalic acid (pH 4.0) and ethyl acetate. The samples were cleaned up by solid phase extraction (SPE) procedure using, at first carboxylic acid and then polymeric Strata X cartridges. Chromatographic separation of DC by LC-UV was achieved on a Luna C8 analytical column and for LC-MS/MS analysis Luna C18 column was used. The presented procedures were evaluated according to the Commission Decision 2002/657/EC. Specificity, decision limit (CCα), detection capacity (CCβ), recovery (absolute and relative), precision (repeatability and reproducibility) were determined during validation process. The limit of detection (LOD) was 10μg/kg for LC-UV and 1μg/kg for LC-MS/MS method. The limit of quantitation (LOQ) was 15 and 2μg/kg for LC-UV and LC-MS/MS, respectively. The absolute recovery for the LC-UV and relative recovery for the LC-MS/MS method at 300μg/kg concentration level were 79%; 101% for fat and 82%; 99% for fat and skin, respectively. The developed liquid chromatography with ultraviolet detection (LC-UV) and liquid chromatography tandem mass spectrometry (LC-MS/MS) methods have been applied to quantitative determination of doxycycline (DC) in samples of chicken fat tissue obtained from animals treated with DC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Doxycyclin -hyclat
Sigma-Aldrich
Doxycyclinhydrochlorid, gebrauchsfertige Lösung, 100 mg/mL in DMSO, ≥95.0% (HPLC)
Supelco
Doxycyclin -hyclat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxycycline monohydrate
Supelco
Doxycyclin -hyclat, VETRANAL®, analytical standard