Merck
  • Assessment of xanthan gum based sustained release matrix tablets containing highly water-soluble propranolol HCl.
Assessment of xanthan gum based sustained release matrix tablets containing highly water-soluble propranolol HCl.
Acta poloniae pharmaceutica (2013-04-26)
Atif Ali, Muhammad Iqbal, Naveed Akhtar, Haji Muhamad Shoaib Khan, Aftab Ullah, Minhaj Uddin, Muhammad Tahir Khan
ZUSAMMENFASSUNG
The present study was carried out to develop oral sustained release tablets of propranolol HCl by different ratios of drug : matrix. Tablets were prepared by direct compression technique using xanthan gum and lactose. All the formulations (tablets) were evaluated for thickness, diameter, hardness, friability, weight variation, content of active ingredient, in vitro dissolution using USP dissolution apparatus-II and swelling index. In case of dissolution, an inverse relationship was noted between amount of xanthan gum and release rate of propranolol HCl and the drug release was gradually enhanced as the amount of the lactose increased. The direct release was observed between swelling index and xanthan gum concentration. Significant difference in different media was observed in release profile, indicating that propranolol HCI has better solubility in HCI buffer pH 1.2. Moreover, dissolution data at differing stirring speeds was also analyzed, indicating that the drug release profile was at 50 rpm comparative to 100 rpm. The kinetic treatment showed the best fitted different mathematical models (zero order, first order, Higuchi's, Hixson-Crowell and Korsmeyer Peppas model. Most of the formulations showed linearity in Higuchi's model. The drug release from these tablets was by Fickian diffusion and anomalous (non-Fickian) mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-Propranolol -hydrochlorid, ≥99% (TLC), powder
Supelco
Lactose, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Supelco
Propranolol-Hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Lactose, wasserfrei, European Pharmacopoeia (EP) Reference Standard
Supelco
Propranolol -hydrochlorid, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Propranolol -hydrochlorid, European Pharmacopoeia (EP) Reference Standard