Merck
Management of drug induced sexual dysfunction in male rats by ethyl acetate fraction of onion.
Acta poloniae pharmaceutica (2013-04-26)
Neelesh Malviya, Sanjay Jain, Vipin Bihari Gupta, Savita Vyas
ZUSAMMENFASSUNG
The present study aimed to investigate the effect of ethyl acetate fraction of A. cepa bulb on mating behavior in paroxetine-induced sexually dysfunction male rats. Sexual dysfunctions such as decreased libido, delayed orgasm, difficulties in maintaining an erection, and inhibition of ejaculation are common side effects of paroxetine. A. cepa bulb ethyl acetate fraction (200 mg/kg) was administered orally in paroxetine-induced sexually impaired male rats for 7 days. At the end of 7th day, mount frequency (MF), intromission frequency (IF), ejaculatory frequency (EF), mount latency (ML), intromission latency (IL), ejaculatory latency (EL) and post-ejaculatory interval (PEI) were the parameters observed. Results showed that in relation to the paroxetine treated group, ethyl acetate fraction, significantly restored the normal sexual behavior as evident from increased MF, IF, EF and reduced ML, IL, EL and PEI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Supelco
Ethylacetat, analytical standard
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Paroxetin -hydrochlorid Hemihydrat, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Paroxetin -hydrochlorid Hemihydrat, European Pharmacopoeia (EP) Reference Standard
Paroxetin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard