Merck
The catalytic mechanism for aerobic formation of methane by bacteria.
Nature (2013-04-26)
Siddhesh S Kamat, Howard J Williams, Lawrence J Dangott, Mrinmoy Chakrabarti, Frank M Raushel
ZUSAMMENFASSUNG
Methane is a potent greenhouse gas that is produced in significant quantities by aerobic marine organisms. These bacteria apparently catalyse the formation of methane through the cleavage of the highly unreactive carbon-phosphorus bond in methyl phosphonate (MPn), but the biological or terrestrial source of this compound is unclear. However, the ocean-dwelling bacterium Nitrosopumilus maritimus catalyses the biosynthesis of MPn from 2-hydroxyethyl phosphonate and the bacterial C-P lyase complex is known to convert MPn to methane. In addition to MPn, the bacterial C-P lyase complex catalyses C-P bond cleavage of many alkyl phosphonates when the environmental concentration of phosphate is low. PhnJ from the C-P lyase complex catalyses an unprecedented C-P bond cleavage reaction of ribose-1-phosphonate-5-phosphate to methane and ribose-1,2-cyclic-phosphate-5-phosphate. This reaction requires a redox-active [4Fe-4S]-cluster and S-adenosyl-L-methionine, which is reductively cleaved to L-methionine and 5'-deoxyadenosine. Here we show that PhnJ is a novel radical S-adenosyl-L-methionine enzyme that catalyses C-P bond cleavage through the initial formation of a 5'-deoxyadenosyl radical and two protein-based radicals localized at Gly 32 and Cys 272. During this transformation, the pro-R hydrogen from Gly 32 is transferred to the 5'-deoxyadenosyl radical to form 5'-deoxyadenosine and the pro-S hydrogen is transferred to the radical intermediate that ultimately generates methane. A comprehensive reaction mechanism is proposed for cleavage of the C-P bond by the C-P lyase complex that uses a covalent thiophosphate intermediate for methane and phosphate formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycin -hydrochlorid, ≥99% (HPLC)
SAFC
L-Methionin
Sigma-Aldrich
Tributylzinnhydrid, contains 0.05% BHT as stabilizer, 97%
Sigma-Aldrich
L-Methionin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionin, reagent grade, ≥98% (HPLC)
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Methionin, ≥99%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Supelco
L-Methionin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Sigma-Aldrich
Glycin 1 M -Lösung
Methionin, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
DL-Methionin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionin, ≥99.0% (NT)
Supelco
Glycin -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Methionin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.
Sigma-Aldrich
[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11, ≥97% (HPLC)