Merck
Imaging: Magnetic bacteria on a diamond plate.
Nature (2013-04-27)
Mihály Pósfai, Rafal E Dunin-Borkowski
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diamant, nanopowder, <10 nm particle size (TEM), ≥97% trace metals basis
Sigma-Aldrich
Diamant, nanopowder, <10 nm particle size (TEM), ≥95% trace metals basis
Sigma-Aldrich
Diamant, synthetic monocrystalline powder, ≤1 μm