Merck
  • Functional characterization of an abiotic stress-inducible transcription factor AtERF53 in Arabidopsis thaliana.
Functional characterization of an abiotic stress-inducible transcription factor AtERF53 in Arabidopsis thaliana.
Plant molecular biology (2013-04-30)
En-Jung Hsieh, Mei-Chun Cheng, Tsan-Piao Lin
ZUSAMMENFASSUNG
AP2/ERF proteins play crucial roles in plant growth and development and in responses to biotic and abiotic stresses. ETHYLENE RESPONSE FACTOR 53 (AtERF53) belongs to group 1 in the ERF family and is induced in the early hours of dehydration and salt treatment. The functional study of AtERF53 is hampered because its protein expression in Arabidopsis is vulnerable to degradation in overexpressed transgenic lines. Taking advantage of the RING domain ligase1/RING domain ligase2 (rglg1rglg2) double mutant in which the AtERF53 can express stably, we investigate the physiological function of AtERF53. In this study, we demonstrate that expression of AtERF53 in wild-type Arabidopsis was responsive to heat and abscisic acid (ABA) treatment. From results of the cotransfection experiment, we concluded that AtERF53 has positive transactivation activity. Overexpression of AtERF53 in the rglg1rglg2 double mutant conferred better heat-stress tolerance and had resulted in higher endogenous ABA and proline levels compared to rglg1rglg2 double mutants. AtERF53 also has a function to regulate guard-cell movement because the stomatal aperture of AtERF53 overexpressed in rglg1rglg2 double mutant was smaller than that in the rglg1rglg2 double mutant under ABA treatment. In a global gene expression study, we found higher expressions of many stress-related genes, such as DREB1A, COR15A, COR15B, PLC, P5CS1, cpHSC70 s and proline and ABA metabolic-related genes. Furthermore, we identified several downstream target genes of AtERF53 by chromatin immunoprecipitation assay. In conclusion, the genetic, molecular and biochemical result might explain how AtERF53 serving as a transcription factor contributes to abiotic stress tolerance in Arabidopsis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Prolin
Sigma-Aldrich
L-Prolin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
2-cis,4-trans-Abscisinsäure, synthetic, 98%
Sigma-Aldrich
L-Prolin, 99%, FCC, FG
Sigma-Aldrich
(+)-Abscisinsäure, ≥98% (HPLC)
Supelco
L-Prolin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland