Merck
Zirconia implants supporting overdentures: a pilot study with novel prosthodontic designs.
The International journal of prosthodontics (2013-04-30)
Reham B Osman, Alan G T Payne, Warwick Duncan, Sunyoung Ma
ZUSAMMENFASSUNG
Limited clinical research identifies prosthodontic perspectives of novel designs for zirconia implants supporting overdentures. Four pilot study participants were selected before a planned randomized clinical trial on zirconia implants supporting overdentures. Novel designs for maxillary four-implant overdentures (quadrilateral design) and mandibular three-implant overdentures (tripodal design) were used with 28 implants (maxilla, n = 16; mandible, n = 12). Four implants failed to achieve osseointegration prior to loading. At the 1-year follow-up appointment, all implants were surviving, the overdentures were in function, and there were no clinical signs of wear of the attachment system. A proof-of-principle for prosthodontic perspectives of a novel design using one-piece zirconia implants supporting maxillary and mandibular implant overdentures was achieved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zirkonium(IV)-oxid, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirkonium(IV)-oxid, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirkonium(IV)-oxid, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirkonium(IV)-oxid -Lösung, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirkonium(IV)-oxid -Lösung, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O