Merck
Judge: No age restriction for emergency contraceptive.
JAMA (2013-05-02)
Bridget M Kuehn
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
D(−)-Norgestrel, analytical standard
Levonorgestrel, European Pharmacopoeia (EP) Reference Standard