Merck
Pyridinylquinazolines selectively inhibit human methionine aminopeptidase-1 in cells.
Journal of medicinal chemistry (2013-05-03)
Feiran Zhang, Shridhar Bhat, Sandra B Gabelli, Xiaochun Chen, Michelle S Miller, Benjamin A Nacev, Yim Ling Cheng, David J Meyers, Karen Tenney, Joong Sup Shim, Phillip Crews, L Mario Amzel, Dawei Ma, Jun O Liu
ZUSAMMENFASSUNG
Methionine aminopeptidases (MetAPs), which remove the initiator methionine from nascent peptides, are essential in all organisms. While MetAP2 has been demonstrated to be a therapeutic target for inhibiting angiogenesis in mammals, MetAP1 seems to be vital for cell proliferation. Our earlier efforts identified two structural classes of human MetAP1 (HsMetAP1)-selective inhibitors (1-4), but all of them failed to inhibit cellular HsMetAP1. Using Mn(II) or Zn(II) to activate HsMetAP1, we found that 1-4 could only effectively inhibit purified HsMetAP1 in the presence of physiologically unachievable concentrations of Co(II). In an effort to seek Co(II)-independent inhibitors, a novel structural class containing a 2-(pyridin-2-yl)quinazoline core has been discovered. Many compounds in this class potently and selectively inhibited HsMetAP1 without Co(II). Subsequently, we demonstrated that 11j, an auxiliary metal-dependent inhibitor, effectively inhibited HsMetAP1 in primary cells. This is the first report that an HsMetAP1-selective inhibitor is effective against its target in cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidin, ≥99%
SAFC
L-Methionin
Sigma-Aldrich
L-Methionin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Sigma-Aldrich
DL-Methionin, ≥99%
Sigma-Aldrich
Kobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Kobalt, powder, <150 μm, ≥99.9% trace metals basis
Supelco
L-Methionin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Sigma-Aldrich
Thymidin, ≥99.0% (HPLC)
Sigma-Aldrich
Mangan, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Kobalt, granular, 99.99% trace metals basis
Methionin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Kobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
DL-Methionin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionin, ≥99.0% (NT)
Sigma-Aldrich
Kobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Supelco
L-Methionin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Kobalt, rod, diam. 5.0 mm, 99.95% trace metals basis