Merck
A 30-year-old woman with recurrent lower respiratory tract infections.
Chest (2013-05-08)
Faisal A Khasawneh, Adeline J Jou-Tindo
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Levofloxacin, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ofloxacin, VETRANAL®, analytical standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard