Merck
  • [Gender-dependent effects of histone deacetylase inhibitor sodium valproate on early olfactory learning in 129Sv mice].
[Gender-dependent effects of histone deacetylase inhibitor sodium valproate on early olfactory learning in 129Sv mice].
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova (2013-05-09)
O V Burenkova, E A Aleksandrova, I Iu Zaraĭskaia
ZUSAMMENFASSUNG
In the brain, histone acetylation underlies both learning and the maintenance of long-term sustained effects of early experience which is further epigenetically inherited. However, the role of acetylation in learning previously has only been studied in adult animals: high level of learning could be dependent on high levels of histone H3 acetylation in the brain. The role of acetylation in the mechanisms of early learning has not been studied. In the present work, we were interested whether histone deacetylase inhibitor sodium valproate which increases the level of histone H3 acetylation will affect early olfactory discrimination learning in 8-day-old pups of 129Sv mice that are characterized by low efficiency of learning with imitation of maternal grooming. Multiple valproate injections from 3rd to 6th postnatal day had a gender-dependent effect: learning was selectively improved in male but not in female pups. In the female pups, learning improvement was observed after multiple injections of saline. Possible epigenetic mechanisms underlying these sex differences are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Valproinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentansäure
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®