Merck
[Determination of traces of gallium in air by oscillographic polarography].
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao (1990-03-01)
C Sun, Z Yang, D Cheng, Z Yi, T Shen, Z Liu, Y Bai
ZUSAMMENFASSUNG
Gallium and its compounds in air collected by millipore filter membrane are leached by 10% HNO3 at room temperature. After being adjusted with ammonia solution to pH8-9, the leachate is filtered and the filtrate is evaporated to dryness on a water bath. The residue is dissolved with 2 ml of 10% HNO3 and then into the solution are added 0.5 ml of 5% hydroxylamine hydrochloride solution, 0.20 ml of 5% sulfosalicyclic acid solution, 0.5 ml of NaF solution (1mg F/ml) and 1 drop of methyl red solution. Let it stand for 5 min after the color of the solution turns orange by addition of 1:1 NH4OH solution. Add 0.20ml of 0.20% cupferron solution, 0.20ml of 0.5% diphenyl-guanidine solution (prepared in 95% ethanol) and 0.5ml of acetic acid-ammonium acetate buffer (pH 7.0). Then add water to make the final volume of the test solution to be 10ml and measure the height of the second derivative polarographic wave at -1.08V (vs.SCE). The sensitivity of the method is 0.005-0.10 microgram/ml, the coefficient of variation 6.8%, recovery 97.8%. Within 0.005-0.10 microgram/ml, the wave height is linear with the concentration of gallium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Diphenylguanidin, 97%
Supelco
1,3-Diphenylguanidin, analytical standard (for titrimetry)