Merck
Respirable quartz exposure on two medium-sized farms in southern Mozambique.
International journal of occupational and environmental health (2013-05-21)
José C Franque Mirembo, Andrew J Swanepoel, David Rees
ZUSAMMENFASSUNG
Little is known about the potential for overexposure to respirable quartz in farming, in most parts of the world. To measure respirable dust and quartz exposure of tractor operators on two medium-sized dry climate farms. This is a descriptive cross-sectional study of dust exposure of four tractor operators. Farms were selected by convenience sampling. The MDHS 14/3 and FTIR MDHS 101 HSE methods were used to measure dust and to analyze the mass of quartz in dust, respectively. Seventy respirable dust measurements were done. Respirable dust and quartz ranged from 0·01 to 2·88 and 0·001 to 0·30 mg/m(3), respectively. All operators had at least one respirable quartz exposure above 0·1 mg/m(3). Only 17% of respirable quartz concentrations were lower than the ACGIH TLV of 0·025 mg/m(3). The potential for overexposure to respirable quartz was demonstrated. There was a great deal of exposure variability on these farms which has implications for sampling strategies for dust in farming.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quartz, 50-70 mesh particle size