Merck
Radiation treatments after breast-conserving therapy for elderly patients.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-05-22)
Benjamin D Smith, Thomas A Buchholz
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen -citrat (Salz), ≥99%
Tamoxifencitrat für Performance-Test, European Pharmacopoeia (EP) Reference Standard
Tamoxifencitrat, European Pharmacopoeia (EP) Reference Standard