Merck
  • Antiapoptotic effects of cerium oxide and yttrium oxide nanoparticles in isolated rat pancreatic islets.
Antiapoptotic effects of cerium oxide and yttrium oxide nanoparticles in isolated rat pancreatic islets.
Human & experimental toxicology (2013-05-23)
A Hosseini, M Baeeri, M Rahimifard, M Navaei-Nigjeh, A Mohammadirad, N Pourkhalili, S Hassani, M Kamali, M Abdollahi
ZUSAMMENFASSUNG
Type I diabetes mellitus is a metabolic disease caused by the impairment of pancreatic β-cells mainly mediated through oxidative stress and related apoptosis. Islets transplantation seems a promising treatment for these patients, but during islets transplant, various types of stresses related to the isolation and transplantation procedure compromise the function and viability of islets. We recently hypothesized that the combination of cerium oxide (CeO2) and yttrium oxide (Y2O3) nanoparticles with a potential free radical scavenger behavior should be useful to make isolated islets survive until transplanted. In the present study, oxidative stress-induced apoptosis in isolated rat pancreatic islets exposed to hydrogen peroxide (H2O2) and the protective effects of CeO2 and Y2O3 nanoparticles were investigated. Exposure of islets to H2O2 (50 µm, 2 h) increased intracellular oxidant formation such as reactive oxygen species and subsequently apoptosis and decreased viability, glucose-induced adenosine triphosphate (ATP) production and glucose-stimulated insulin secretion. Pretreatment with CeO2 and/or Y2O3 nanoparticles reduced the oxidant formation and apoptosis and increased viability, glucose-induced ATP production and glucose-stimulated insulin secretion. These results suggest that this combination may protect β-cell apoptosis by improving the oxidative stress-mediated apoptotic pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cer(IV)-oxid, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Yttrium(III)-oxid, 99.99% trace metals basis
Sigma-Aldrich
Cer(IV)-oxid, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Cer(IV)-oxid, ≥99.0%
Sigma-Aldrich
Cer(IV)-oxid, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Yttrium(III)-oxid, nanopowder, <50 nm particle size
Sigma-Aldrich
Cerium(IV)-oxid, Dispersion -Lösung, nanoparticles, <25 nm particle size, 10 wt. % in H2O
Sigma-Aldrich
Cer(IV)-oxid, powder, 99.995% trace metals basis
Sigma-Aldrich
Yttrium(III)-oxid, 99.999% trace metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cer, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV)-oxid, Dispersion -Lösung, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Cer, chips
Sigma-Aldrich
Yttrium-Sputtering-Target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Sigma-Aldrich
Yttrium(III)-oxid, Dispersion, 10 wt. % in isopropanol, nanoparticles, <100 nm (DLS), ≥99.9% trace metals basis