Merck
  • Impact of a shortage of first-line antituberculosis medication on tuberculosis control - United States, 2012-2013.
Impact of a shortage of first-line antituberculosis medication on tuberculosis control - United States, 2012-2013.
MMWR. Morbidity and mortality weekly report (2013-05-24)
ZUSAMMENFASSUNG
Tuberculosis (TB) disease is treated in most cases with a regimen of several drugs taken for 6-9 months. Currently, 10 drugs are approved by the Food and Drug Administration (FDA) for treatment of TB. Of these, the four drugs that form the core for first-line treatment regimens are isoniazid (INH), rifampin, ethambutol, and pyrazinamide. In November 2012, the United States began to experience a severe interruption in the supply of INH. To assess the extent of the problem and its impact on TB control programs, a nationwide survey of programs was conducted in January 2013 by the National Tuberculosis Controllers Association (NTCA). The results indicated that the INH shortage was interfering with patient care and could contribute to TB transmission in the United States. This report summarizes the findings of that survey, which showed that 79% of the responding health departments reported difficulties with procuring INH within the last month, with 15% reporting that they no longer had INH and 41% reporting that they would no longer have a supply within 1 month of the survey. Because of local interruptions in INH supply, responding TB programs were changing INH suppliers (69%), prioritizing patients for treatment of latent TB infection (LTBI) (72%), delaying LTBI treatment (68%), and changing to alternative LTBI treatment regimens (88%). Potential solutions for alleviating the INH shortage and averting future shortages include maintaining a national supply of first-line drugs, sharing INH among jurisdictions, working with the World Health Organization's Global Drug Facility to obtain INH from foreign manufacturers, and strengthening reporting of shortages and impending shortages by drug suppliers to FDA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Isoniazid, analytical standard, ≥99% (TLC)