Merck
Tramadol as a local anesthetic: prospects for use in oncological day surgery.
Plastic and reconstructive surgery (2013-05-30)
Maria B Passavanti, Elisa Grella, Maria C Pace, Terasa L Di Gennaro, Caterina Aurilio