Merck
  • Antioxidant responses and renal crystal formation in rainbow trout treated with melamine administered individually or in combination with cyanuric acid.
Antioxidant responses and renal crystal formation in rainbow trout treated with melamine administered individually or in combination with cyanuric acid.
Journal of toxicology and environmental health. Part A (2013-06-01)
Nicole Pacini, Marino Prearo, Maria Cesarina Abete, Paola Brizio, Ambrosius Josef Martin Dörr, Renate Reimschuessel, Wendy Andersen, Laura Gasco, Marzia Righetti, Antonia Concetta Elia
ZUSAMMENFASSUNG
In 2007 and 2008, renal stone formation and kidney damage in human infants were linked to consumption of melamine (MEL)-contaminated infant formula, as well as renal failure and death in pets due to pet food containing both MEL and cyanuric acid (CYA). The aim of this study was to examine the effects of MEL and CYA administered individually or in combination on concentrations of certain metabolites and enzyme activities that serve as markers for oxidative stress in kidney and liver of rainbow trout. In addition, the levels of muscle MEL and renal crystal formation were determined. Trout were fed MEL and/or CYA for 8 wk at 250, 500, or 1000 mg of each compound/kg in feed. Fish muscle residues of MEL exhibited a dose-response relationship. Coexposure of trout to MEL and CYA at the highest dose led to lower MEL residue concentrations in muscle compared to exposure to MEL alone. Renal MEL-CYA complexes were found in kidneys of fish treated with combined MEL and CYA. A dose response was evident with respect to both (1) number of trout displaying renal crystals and (2) number of crystals per fish. Changes in concentration of antioxidant parameters, such as glutathione, superoxide dismutase, catalase, glutathione peroxidase, and glutathione S-transferase, were recorded in both tissues of MEL- and CYA-dosed trout. Lipid peroxidation was more pronounced in kidney than liver. Therefore, feed contaminated with both MEL and CYA could be problematic for fish, as MEL administered to trout, individually or in combination with CYA, may facilitate the onset of oxidative damage in trout.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Melamin, 99%
Sigma-Aldrich
Cyanursäure, 98%
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Supelco
Melamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melamin, analytical standard
Supelco
Cyanursäure, analytical standard