Merck
  • Unique oligosaccharide (apparently glucotetrasaccharide) in urine of patients with glycogen storage diseases.
Unique oligosaccharide (apparently glucotetrasaccharide) in urine of patients with glycogen storage diseases.
Clinical chemistry (1990-07-01)
K Oberholzer, A C Sewell
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glucose-tetrasaccharid, ≥99.0% (TLC)